2,3,5,6-Tetrabrom-p-xylol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2,4,5-Tetrabromo-3,6-dimethylbenzene
OR51854-100G
OR51854-25G
OR51854-5G
OR51854-500G
APOSOR51854-100GEA
371
EUR
APOSOR51854-100G
APOSOR51854-25G
APOSOR51854-5G
APOSOR51854-500G
2,3,5,6-Tetrabrom-p-xylol
2,3,5,6-Tetrabrom-p-xylol
Formel:
C₈H₆Br₄ Molecular Weight: 421.75 g/mol Schmelzpunkt: 254…256 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010353 CAS-Nummer: 23488-38-2 EINECS: 245-688-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...