2-Iodo-6-methylbenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR51857-1G OR51857-250MG OR51857-5G
APOSOR51857-1GEA 0 EUR
APOSOR51857-1G APOSOR51857-250MG APOSOR51857-5G
2-Iodo-6-methylbenzyl bromide
2-Iodo-6-methylbenzyl bromide
Formel: C₈H₈BrI
Lagertemperatur: Raumtemperatur
MDL: MFCD18397826
CAS-Nummer: 1261481-48-4
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR