2-Bromo-4-methoxybenzoic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR51862-1G
OR51862-5G
OR51862-25G
APOSOR51862-1GEA
21.9
EUR
APOSOR51862-1G
APOSOR51862-5G
APOSOR51862-25G
2-Bromo-4-methoxybenzoic acid 98%
2-Bromo-4-methoxybenzoic acid
Formel:
C₈H₇BrO₃ Molecular Weight: 231.0434 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044843 CAS-Nummer: 74317-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...