N-Boc-D-valin 99%

Lieferant: Apollo Scientific

Synonyme: Boc-D-Val-OH

OR51864-100G
APOSOR51864-100GEA 55.1 EUR
APOSOR51864-100G
N-Boc-D-valin 99%
N-Boc-D-valin
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217,3 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038282
CAS-Nummer: 22838-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR