Dibenzyl sulphone

Lieferant: Apollo Scientific

Synonyme: (sulphonylbis(methylene))dibenzene, Benzyl sulphone

OR51866-10G
APOSOR51866-10GEA 139 EUR
APOSOR51866-10G
Dibenzyl sulphone
Dibenzyl sulphone
Formel: C₁₄H₁₄O₂S
Molecular Weight: 246,3248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00022036
CAS-Nummer: 620-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR