2-Bromo-1-(3,4-dimethoxyphenyl)ethanone

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,4-Dimethoxyphenacyl bromide

OR51867-1G OR51867-5G OR51867-10G
APOSOR51867-1GEA 21.9 EUR
APOSOR51867-1G APOSOR51867-5G APOSOR51867-10G
2-Bromo-1-(3,4-dimethoxyphenyl)ethanone
2-Bromo-1-(3,4-dimethoxyphenyl)ethanone
Formel: C₁₀H₁₁BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03425186
CAS-Nummer: 1835-02-5
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR