2-Bromo-1-(3,4-dimethoxyphenyl)ethanone
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,4-Dimethoxyphenacyl bromide
OR51867-1G
OR51867-5G
OR51867-10G
APOSOR51867-1GEA
21.9
EUR
APOSOR51867-1G
APOSOR51867-5G
APOSOR51867-10G
2-Bromo-1-(3,4-dimethoxyphenyl)ethanone
2-Bromo-1-(3,4-dimethoxyphenyl)ethanone
Formel:
C₁₀H₁₁BrO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425186 CAS-Nummer: 1835-02-5 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...