8-Bromo-5-(methylthio)[1,2,4]triazolo[4,3-c]pyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Bromo-5-(methylthio)-[1,2,4]triazolo[4,3-c]pyrimidine, 8-Bromo-5-(methylsulphanyl)[1,2,4]triazolo[4,3-c]pyrimidine

OR51877-1G
APOSOR51877-1GEA 213 EUR
APOSOR51877-1G
8-Bromo-5-(methylthio)[1,2,4]triazolo[4,3-c]pyrimidine
8-Bromo-5-(methylthio)[1,2,4]triazolo[4,3-c]pyrimidine
Formel: C₆H₅BrN₄S
Molecular Weight: 245,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28502491
CAS-Nummer: 99451-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR