(+)-Benzotetramisole 98+%
Lieferant: Apollo Scientific
Synonyme:
(R)-2-Phenyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole, (+)-BTM, (2R)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole
OR51900-1G
APOSOR51900-1GEA
116
EUR
APOSOR51900-1G
(+)-Benzotetramisole 98+%
(+)-Benzotetramisole
Organocatalyst for the kinetic resolution of secondary alcohols by acylation with anhydrides.
Formel:
C₁₅H₁₂N₂S Molecular Weight: 252,34 g/mol Schmelzpunkt: 93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11112169 CAS-Nummer: 885051-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...