3,5-Diamino-6-chloropyrazine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR51902-1G
APOSOR51902-1GEA 575 EUR
APOSOR51902-1G
3,5-Diamino-6-chloropyrazine-2-carboxylic acid
3,5-Diamino-6-chloropyrazine-2-carboxylic acid
Formel: C₅H₅ClN₄O₂
Molecular Weight: 188,572 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208349
CAS-Nummer: 4878-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR