cis-4-Aminocyclohexanol hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: cis-4-Aminocyclohexan-1-ol hydrochloride, cis-4-Hydroxycyclohexylamine hydrochloride

OR51931-5G
APOSOR51931-5GEA 81.2 EUR
APOSOR51931-5G
cis-4-Aminocyclohexanol hydrochloride 97%
cis-4-Aminocyclohexanol hydrochloride
Formel: C₆H₁₄ClNO
Molecular Weight: 151.63 g/mol
Schmelzpunkt: 193…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07366531
CAS-Nummer: 56239-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR