4-(6-Amino-3-pyridyl)-1-boc-piperazine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-4-(6-amino-3-pyridyl)piperazine, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester
OR51957-100G
APOSOR51957-100GEA
63.8
EUR
APOSOR51957-100G
4-(6-Amino-3-pyridyl)-1-boc-piperazine 98%
4-(6-Amino-3-pyridyl)-1-boc-piperazine
Formel:
C₁₄H₂₂N₂O₄ Molecular Weight: 278.35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11594962 CAS-Nummer: 571188-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...