1-(2,4-Dichloropyrimidin-5-yl)ethan-1-one

Lieferant: Apollo Scientific
Achtung

OR51968-1G
APOSOR51968-1GEA 1130 EUR
APOSOR51968-1G
1-(2,4-Dichloropyrimidin-5-yl)ethan-1-one
1-(2,4-Dichloropyrimidin-5-yl)ethan-1-one
Formel: C₆H₄Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09754024
CAS-Nummer: 871254-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR