2-Brom-5-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR51974-1G OR51974-25G OR51974-5G
APOSOR51974-1GEA 21.9 EUR
APOSOR51974-1G APOSOR51974-25G APOSOR51974-5G
2-Brom-5-nitrophenol
2-Brom-5-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218.0100 g/mol
Schmelzpunkt: 116…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00466299
CAS-Nummer: 52427-05-1
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR