Ethyl-5-bromo-2-methylnicotinate 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-bromo-2-methylnicotinate
OR51975-100G
OR51975-25G
OR51975-5G
APOSOR51975-100GEA
1860
EUR
APOSOR51975-100G
APOSOR51975-25G
APOSOR51975-5G
Ethyl-5-bromo-2-methylnicotinate 97+%
Ethyl-5-bromo-2-methylnicotinate
Formel:
C₉H₁₀BrNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698571 CAS-Nummer: 129477-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...