Isonicotinaldehydoxim 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Pyridinecarbaldehyde oxime, Pyridine-4-carboxaldoxime

OR51992-100G
APOSOR51992-100GEA 166 EUR
APOSOR51992-100G
Isonicotinaldehydoxim 98%
Isonicotinaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122,13 g/mol
Schmelzpunkt: 130…133 °C
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006446
CAS-Nummer: 696-54-8
EINECS: 271-796-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR