4-Brom-4'-methoxybiphenyl 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-4'-methoxy-1,1'-biphenyl, 1-Bromo-4-(4-methoxyphenyl)benzene
OR52012-5G
APOSOR52012-5GEA
55.1
EUR
APOSOR52012-5G
4-Brom-4'-methoxybiphenyl 98%
4-Brom-4'-methoxybiphenyl
Formel:
C₁₃H₁₁BrO Molecular Weight: 263,1298 g/mol Schmelzpunkt: 143…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671954 CAS-Nummer: 58743-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...