2-Amino-4-brombenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR52035-1G
APOSOR52035-1GEA 14.7 EUR
APOSOR52035-1G
2-Amino-4-brombenzonitril 98%
2-Amino-4-brombenzonitril
Formel: C₇H₅BrN₂
Molecular Weight: 197,0300 g/mol
Schmelzpunkt: 162…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08752638
CAS-Nummer: 304858-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR