3-Amino-1H-pyrazolo[3,4-c]pyridine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Pyrazolo[3,4-c]pyridin-3-amine, 3-Amino-6-azaindole

OR52041-100MG
APOSOR52041-100MGEA 104 EUR
APOSOR52041-100MG
3-Amino-1H-pyrazolo[3,4-c]pyridine 95+%
3-Amino-1H-pyrazolo[3,4-c]pyridine
Formel: C₆H₆N₄
Molecular Weight: 134,139 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11656373
CAS-Nummer: 76006-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR