3-Brom-2-hydroxybenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR52048-1G
OR52048-5G
APOSOR52048-1GEA
24.8
EUR
APOSOR52048-1G
APOSOR52048-5G
3-Brom-2-hydroxybenzonitril
3-Brom-2-hydroxybenzonitril
Formel:
C₇H₄BrNO Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11042854 CAS-Nummer: 13073-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...