3'-Bromo-2'-methoxyacetophenone 97%

Lieferant: Apollo Scientific
Achtung

OR52056-1G OR52056-250MG OR52056-5G
APOSOR52056-1GEA 29 EUR
APOSOR52056-1G APOSOR52056-250MG APOSOR52056-5G
3'-Bromo-2'-methoxyacetophenone 97%
3'-Bromo-2'-methoxyacetophenone
Formel: C₉H₉BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22573042
CAS-Nummer: 267651-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR