Methyl 4-[(1S)-1-aminoethyl]benzoate 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (S)-4-(Methoxycarbonyl)-α-methylbenzylamine, (S)-Methyl 4-(1-aminoethyl)benzoate, (1S)-1-[4-(Methoxycarbonyl)phenyl]ethylamine

OR52064-5G
APOSOR52064-5GEA 68.3 EUR
APOSOR52064-5G
Methyl 4-[(1S)-1-aminoethyl]benzoate 98%
Methyl 4-[(1S)-1-aminoethyl]benzoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Schmelzpunkt: 59…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06762070
CAS-Nummer: 222714-37-6
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR