6-Ethylpyrimidine-2,4-diamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Diamino-6-ethylpyrimidine, 2,4-Diamino-6-ethyl-1,3-diazine

OR52065-1G
APOSOR52065-1GEA 66.7 EUR
APOSOR52065-1G
6-Ethylpyrimidine-2,4-diamine 97%
6-Ethylpyrimidine-2,4-diamine
Formel: C₆H₁₀N₄
Molecular Weight: 138,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02091119
CAS-Nummer: 514854-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR