2-Bromoethylamine, N-CBZ protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Amino-2-bromoethane, N-CBZ protected, Benzyl (2-bromoethyl)carbamate

OR52103-100G
APOSOR52103-100GEA 171 EUR
APOSOR52103-100G
2-Bromoethylamine, N-CBZ protected
2-Bromoethylamine, N-CBZ protected
Formel: C₁₀H₁₂BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09909490
CAS-Nummer: 53844-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR