Methylimidazo[1,2-a]pyridin-7-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR52106-1G
APOSOR52106-1GEA
43.5
EUR
APOSOR52106-1G
Methylimidazo[1,2-a]pyridin-7-carboxylat
Methylimidazo[1,2-a]pyridin-7-carboxylat
Formel:
C₉H₈N₂O₂ Schmelzpunkt: 133…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09910350 CAS-Nummer: 86718-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...