5-Chloro-3-(2-chlorobenzyl)-1,2,4-thiadiazole

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-2-[(5-chloro-1,2,4-thiadiazol-3-yl)methyl]benzene, 5-Chloro-3-[(2-chlorophenyl)methyl]-1,2,4-thiadiazole

OR52112-1G
APOSOR52112-1GEA 517 EUR
APOSOR52112-1G
5-Chloro-3-(2-chlorobenzyl)-1,2,4-thiadiazole
5-Chloro-3-(2-chlorobenzyl)-1,2,4-thiadiazole
Formel: C₉H₆Cl₂N₂S
Molecular Weight: 245,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14707311
CAS-Nummer: 90418-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR