5-Chloro-3-[(thien-2-yl)methyl]-1,2,4-thiadiazole

Lieferant: Apollo Scientific

Synonyme: 2-[(5-Chloro-1,2,4-thiadiazol-3-yl)methyl]thiophene

OR52113-1G
APOSOR52113-1GEA 542 EUR
APOSOR52113-1G
5-Chloro-3-[(thien-2-yl)methyl]-1,2,4-thiadiazole
5-Chloro-3-[(thien-2-yl)methyl]-1,2,4-thiadiazole
Formel: C₇H₅ClN₂S₂
Molecular Weight: 216,711 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20257108
CAS-Nummer: 1338218-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR