5-Chloro-3-cyclopropyl-1,2,4-thiadiazole

Lieferant: Apollo Scientific

Synonyme: (5-Chloro-1,2,4-thiadiazol-3-yl]cyclopropane

OR52114-1G
APOSOR52114-1GEA 542 EUR
APOSOR52114-1G
5-Chloro-3-cyclopropyl-1,2,4-thiadiazole
5-Chloro-3-cyclopropyl-1,2,4-thiadiazole
Formel: C₅H₅ClN₂S
Molecular Weight: 160,625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14707316
CAS-Nummer: 122684-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR