5-Chloro-3-(2,3-dimethoxyphenyl)-1,2,4-thiadiazole

Lieferant: Apollo Scientific

Synonyme: 3-(5-Chloro-1,2,4-thiadiazol-3-yl)catechol dimethyl ether, 1-(5-Chloro-1,2,4-thiadiazol-3-yl)-2,3-dimethoxybenzene

OR52115-1G
APOSOR52115-1GEA 542 EUR
APOSOR52115-1G
5-Chloro-3-(2,3-dimethoxyphenyl)-1,2,4-thiadiazole
5-Chloro-3-(2,3-dimethoxyphenyl)-1,2,4-thiadiazole
Formel: C₁₀H₉ClN₂O₂S
Molecular Weight: 256,709 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22373774
CAS-Nummer: 1378261-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR