3-(4-Bromobenzyl)-5-chloro-1,2,4-thiadiazole

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-4-[(5-chloro-1,2,4-thiadiazol-3-yl)methyl]benzene, 3-[(4-Bromophenyl)methyl]-5-chloro-1,2,4-thiadiazole

OR52116-1G
APOSOR52116-1GEA 451 EUR
APOSOR52116-1G
3-(4-Bromobenzyl)-5-chloro-1,2,4-thiadiazole
3-(4-Bromobenzyl)-5-chloro-1,2,4-thiadiazole
Formel: C₉H₆BrClN₂S
Molecular Weight: 289,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14707317
CAS-Nummer: 1221342-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR