5-Chloro-3-(3-methoxyphenyl)-1,2,4-thiadiazole
Lieferant: Apollo Scientific
Synonyme:
3-(5-Chloro-1,2,4-thiadiazol-3-yl)anisole
OR52117-1G
APOSOR52117-1GEA
542
EUR
APOSOR52117-1G
5-Chloro-3-(3-methoxyphenyl)-1,2,4-thiadiazole
5-Chloro-3-(3-methoxyphenyl)-1,2,4-thiadiazole
Formel:
C₉H₇ClN₂OS Molecular Weight: 226,683 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14707350 CAS-Nummer: 1221341-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...