6-Brombenzothiazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromobenzo[d]thiazole
OR52121-5G
APOSOR52121-5GEA
69.6
EUR
APOSOR52121-5G
6-Brombenzothiazol 97%
6-Bromobenzothiazole
Formel:
C₇H₄BrNS Molecular Weight: 214.09 g/mol Schmelzpunkt: 52…54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115372 CAS-Nummer: 53218-26-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...