3-Phenyl-1H-pyrazol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole

OR52122-25G
APOSOR52122-25GEA 101 EUR
APOSOR52122-25G
3-Phenyl-1H-pyrazol 98%
3-Phenyl-1H-pyrazole
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
Schmelzpunkt: 75…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159654
CAS-Nummer: 2458-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR