6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-hydroxybenzo[d]thiazole-2-carbonitrile, 2-Cyano-6-hydroxybenzothiazole, 6-hydroxy-1,3-benzothiazole-2-carbonitrile
OR52126-1G
APOSOR52126-1GEA
43.5
EUR
APOSOR52126-1G
6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
6-Hydroxybenzo[d]thiazole-2-carbonitrile
Formel:
C₈H₄N₂OS Molecular Weight: 176,19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00296905 CAS-Nummer: 939-69-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...