3-Iodo-2-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-6-iodonitrobenzene
OR52139-1G
OR52139-250MG
OR52139-5G
APOSOR52139-1GEA
192
EUR
APOSOR52139-1G
APOSOR52139-250MG
APOSOR52139-5G
3-Iodo-2-nitroaniline
3-Iodo-2-nitroaniline
Formel:
C₆H₅IN₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD18397930 CAS-Nummer: 721925-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...