3-Iodo-2-nitroaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-6-iodonitrobenzene

OR52139-1G OR52139-250MG OR52139-5G
APOSOR52139-1GEA 192 EUR
APOSOR52139-1G APOSOR52139-250MG APOSOR52139-5G
3-Iodo-2-nitroaniline
3-Iodo-2-nitroaniline
Formel: C₆H₅IN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD18397930
CAS-Nummer: 721925-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR