4,4'-Bis(2-methoxystyryl)biphenyl
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4-Bis(2-methoxystyryl)biphenyl, 4,4'-Bis[2-(2-methoxyphenyl)vinyl]biphenyl
OR52141-1KG
OR52141-500G
OR52141-100G
APOSOR52141-1KGEA
148
EUR
APOSOR52141-1KG
APOSOR52141-500G
APOSOR52141-100G
4,4'-Bis(2-methoxystyryl)biphenyl
4,4'-Bis(2-methoxystyryl)biphenyl
Optical brightener. Recommended for use in a wide variety of plastics such as PVC and PP. Also for use with artificial leathers.
Formel:
C₃₀H₂₆O₂ Molecular Weight: 418.54 g/mol Schmelzpunkt: 216…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD23160777 CAS-Nummer: 40470-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...