1,4-Bis(2-benzoxazolyl)naphthalene 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,4-Bis(benzo[d]oxazol-2-yl)naphthalene
OR52142-100G
APOSOR52142-100GEA
82.8
EUR
APOSOR52142-100G
1,4-Bis(2-benzoxazolyl)naphthalene 97%
1,4-Bis(2-benzoxazolyl)naphthalene
Optical brightener. Recommended for the use in a wide variety of plastics, especially EVA foamed plastics and rubber plastics, clear lacquers, UV coating and printing inks.
Formel:
C₂₄H₁₄N₂O₂ Molecular Weight: 362.39 g/mol Schmelzpunkt: 210…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00357168 CAS-Nummer: 5089-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...