(-)-Corey lactone

Lieferant: Apollo Scientific
Achtung

Synonyme: (-)-6β-Hydroxymethyl-7α-hydroxy-cis-2-oxabicyclo[3.3.0]octan-3-one, (3aR,4S,5R,6aS)-5-Hydroxy-4-(hydroxymethyl)hexahydro-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)-Corey lactone diol

OR52144-5G
APOSOR52144-5GEA 120 EUR
APOSOR52144-5G
(-)-Corey lactone
(-)-Corey lactone
Formel: C₈H₁₂O₄
Schmelzpunkt: 115…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075234
CAS-Nummer: 32233-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR