2-Chloroquinoline-5-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR52151-5G
APOSOR52151-5GEA 201 EUR
APOSOR52151-5G
2-Chloroquinoline-5-carboxylic acid 98%
2-Chloroquinoline-5-carboxylic acid
Formel: C₁₀H₆ClNO₂
Molecular Weight: 207.6100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226761
CAS-Nummer: 1092287-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR