Ethyl 3-methoxy-4-oxopiperidine-1-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Carbethoxy-3-methoxy-4-piperidone

OR52157-1G
APOSOR52157-1GEA 242 EUR
APOSOR52157-1G
Ethyl 3-methoxy-4-oxopiperidine-1-carboxylate 97%
Ethyl 3-methoxy-4-oxopiperidine-1-carboxylate
Formel: C₉H₁₅NO₄
Molecular Weight: 201,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093811
CAS-Nummer: 83863-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR