3-(1,3-Thiazol-2-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Carboxyphenyl)-1,3-thiazole

OR5216-1G
APOSOR5216-1GEA 357 EUR
APOSOR5216-1G
3-(1,3-Thiazol-2-yl)benzoic acid 97%
3-(1,3-Thiazol-2-yl)benzoic acid
Formel: C₁₀H₇NO₂S
Molecular Weight: 205,233 g/mol
Schmelzpunkt: 160,5…161.5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06797800
CAS-Nummer: 847956-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR