3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-Carboxyphenyl)-1-methyl-1H-pyrazole

OR5217-1G
APOSOR5217-1GEA 329 EUR
APOSOR5217-1G
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid 97%
3-(1-Methyl-1H-pyrazol-3-yl)benzoic acid
Formel: C₁₁H₁₀N₂O₂
Molecular Weight: 202,209 g/mol
Schmelzpunkt: 167…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09065006
CAS-Nummer: 906352-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR