4-Dodecylbenzenesulphonic acid
Lieferant: Apollo Scientific
|
Gefahr
|
OR52171-100G
OR52171-5G
OR52171-25G
APOSOR52171-100GEA
0
EUR
APOSOR52171-100G
APOSOR52171-5G
APOSOR52171-25G
4-Dodecylbenzenesulphonic acid
4-Dodecylbenzenesulphonic acid
Formel:
C₁₈H₃₀O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066677 CAS-Nummer: 121-65-3 UN: 2586 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...