4-Dodecylbenzenesulphonic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: P-Dodecylbenzenesulphonic acid

OR52171-100G OR52171-5G OR52171-25G
APOSOR52171-100GEA 69.6 EUR
APOSOR52171-100G APOSOR52171-5G APOSOR52171-25G
4-Dodecylbenzenesulphonic acid
4-Dodecylbenzenesulphonic acid
Formel: C₁₈H₃₀O₃S
Molecular Weight: 326.49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066677
CAS-Nummer: 121-65-3
UN: 2586
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR