1,3-Oxazole-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR52173-1G OR52173-250MG
APOSOR52173-1GEA 0 EUR
APOSOR52173-1G APOSOR52173-250MG
1,3-Oxazole-5-carbaldehyde
1,3-Oxazole-5-carbaldehyde
Formel: C₄H₃NO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD02179513
CAS-Nummer: 118994-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR