4-Chloropyridine-2,3-diamine 97%

Lieferant: Apollo Scientific

Synonyme: 2,3-Diamino-4-chloropyridine

OR52182-1G
APOSOR52182-1GEA 87 EUR
APOSOR52182-1G
4-Chloropyridine-2,3-diamine 97%
4-Chloropyridine-2,3-diamine
Formel: C₅H₆ClN₃
Molecular Weight: 143,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13181777
CAS-Nummer: 24484-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR