N-Fmoc-D-valin 99%
Lieferant: Apollo Scientific
Synonyme:
Fmoc-D-Val-OH, D-Valine, N-FMOC protected
OR52196-100G
APOSOR52196-100GEA
79.9
EUR
APOSOR52196-100G
N-Fmoc-D-valin 99%
N-Fmoc-D-valin
Formel:
C₂₀H₂₁NO₄ Molecular Weight: 339,39 g/mol Schmelzpunkt: 143…144 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00062953 CAS-Nummer: 84624-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...