3-Amino-2-chlor-6-methylpyridin 96%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-chloro-6-picoline, 5-Amino-6-chloro-2-picoline

OR52200-25G
APOSOR52200-25GEA 108 EUR
APOSOR52200-25G
3-Amino-2-chlor-6-methylpyridin 96%
3-Amino-2-chloro-6-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142.5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095213
CAS-Nummer: 39745-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR