3-Brom-2-chlor-6-methylpyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-2-chlor-6-picolin, 5-Brom-6-chlor-2-picolin, 5-Brom-6-chlor-2-methylpyridin

OR52202-100G
APOSOR52202-100GEA 282 EUR
APOSOR52202-100G
3-Brom-2-chlor-6-methylpyridin 98%
3-Bromo-2-chloro-6-methylpyridine
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095218
CAS-Nummer: 185017-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR